AC5372910880 Accela ChemBio Inc.

2-(Carboxymethoxy)-4-fluorobenzoic Acid

  • MDL number MFCD18651703
  • Cas Number : 1272756-12-3
  • Molecular Weight : 214.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012810&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012810.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart