AC5372910885 Accela ChemBio Inc.

3-(Chloromethyl)pyrazole Hydrochloride

  • MDL number MFCD08234808
  • Cas Number : 69658-97-5
  • Molecular Weight : 153.00984

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012821&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012821.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart