AC5372910900 Accela ChemBio Inc.

6-Methoxy-3-pyridineacetic Acid

  • MDL number MFCD09924951
  • Cas Number : 902130-87-4
  • Molecular Weight : 167.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013002&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY013002.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart