AC5372910912 Accela ChemBio Inc.

2-Cyanothiazole

  • MDL number MFCD09702472
  • Cas Number : 1452-16-0
  • Molecular Weight : 110.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013085&attname=GoodsCode

Safety Information

signalword Warning
pcodes P260-P261-P280-P305+P351+P338-P405-P501A
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY013085.pdf
hstatements H371-H302-H312-H332-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart