AC5372910943 Accela ChemBio Inc.

N-Benzyl-DL-serine Methyl Ester

  • MDL number MFCD18651713
  • Cas Number : 144001-42-3
  • Molecular Weight : 209.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013175&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY013175.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart