AC5372910944 Accela ChemBio Inc.

3-Acetamidopyrazole

  • MDL number MFCD09702442
  • Cas Number : 3553-12-6
  • Molecular Weight : 125.12858

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013182&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY013182.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart