AC5372910952 Accela ChemBio Inc.

3-Amino-N,N-dimethylpiperidine-1-carboxamide

  • MDL number MFCD18651716
  • Cas Number : 1272756-20-3
  • Molecular Weight : 171.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013241&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY013241.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart