AC5372910954 Accela ChemBio Inc.

2-Amino-5-bromo-4-methoxypyrimidine

  • MDL number MFCD08460376
  • Cas Number : 36082-45-8
  • Molecular Weight : 204.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013305&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY013305.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart