AC5372910961 Accela ChemBio Inc.

O-Butylhydroxylamine

  • MDL number MFCD07473528
  • Cas Number : 5622-77-5
  • Molecular Weight : 89.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013356&attname=GoodsCode

Safety Information

signalword Warning
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY013356.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart