AC5372910972 Accela ChemBio Inc.

Nitrocyclobutane

  • MDL number MFCD12963566
  • Cas Number : 2625-41-4
  • Molecular Weight : 101.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013426&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY013426.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart