AC5372910979 Accela ChemBio Inc.

Methyl 4-(4-Boc-1-piperazinyl)-3-chlorobenzoate

  • MDL number MFCD18651754
  • Cas Number : 906559-46-4
  • Molecular Weight : 354.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013465&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY013465.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart