AC5372910986 Accela ChemBio Inc.

N-Methyl-2-(methylsulfonyl)acetamide

  • MDL number MFCD18651755
  • Cas Number : 1272756-05-4
  • Molecular Weight : 151.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013498&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY013498.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart