AC5372910990 Accela ChemBio Inc.

Diethyl 1-Methylimidazole-4,5-dicarboxylate

  • MDL number MFCD18651756
  • Cas Number : 1210-92-0
  • Molecular Weight : 226.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013520&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY013520.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart