AC5372911001 Accela ChemBio Inc.

1-Boc-3-(1-phenylethylamino)-4-piperidinemethanol

  • MDL number MFCD18911736
  • Cas Number : 1305320-69-7
  • Molecular Weight : 334.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013597&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY013597.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart