AC5372911025 Accela ChemBio Inc.

3-(2,6-Difluorophenyl)propionic Acid

  • MDL number MFCD06823978
  • Cas Number : 167683-63-8
  • Molecular Weight : 186.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to off white solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013788&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY013788.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart