AC5372911036 Accela ChemBio Inc.

2-(Chloromethyl)-3-fluoropyridine Hydrochloride

  • MDL number MFCD19442768
  • Cas Number : 149463-07-0
  • Molecular Weight : 182.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013902&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY013902.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart