AC5372911037 Accela ChemBio Inc.

N-[3-(Trifluoromethyl)benzyl]ethylamine

  • MDL number MFCD09935363
  • Cas Number : 14355-04-5
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013928&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY013928.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart