AC5372911040 Accela ChemBio Inc.

[4-Amino-2-(trifluoromethyl)phenyl](4-methyl-1-piperazinyl)methanone

  • MDL number MFCD18911740
  • Cas Number : 853297-04-8
  • Molecular Weight : 287.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013963&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY013963.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart