AC5372911043 Accela ChemBio Inc.

O-[2-(Vinyloxy)ethyl]hydroxylamine

  • MDL number MFCD09951926
  • Cas Number : 391212-29-6
  • Molecular Weight : 103.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014040&attname=GoodsCode

Safety Information

signalword Warning
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY014040.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart