AC5372911046 Accela ChemBio Inc.

4-[4-[[2-(4-Chlorophenyl)-5,5-dimethyl-1-cyclohexenyl]methyl]-1-piperazinyl]benzoic Acid Hydrochloride

  • MDL number MFCD18911741
  • Cas Number : 1305320-63-1
  • Molecular Weight : 475.45048

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014058&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY014058.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart