AC5372911076 Accela ChemBio Inc.

2-Acetyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic Acid

  • MDL number MFCD00718847
  • Cas Number : 143767-54-8
  • Molecular Weight : 219.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014261&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY014261.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart