AC5372911084 Accela ChemBio Inc.

Dimethyl 2,3-Pyridinedicarboxylate

  • MDL number MFCD00186549
  • Cas Number : 605-38-9
  • Molecular Weight : 195.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014299&attname=GoodsCode

Safety Information

signalword Warning
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY014299.pdf
hstatements H315-H318-H335
Availability details for
Quantity
  • Availability
  • Add to Cart