AC5372911088 Accela ChemBio Inc.

1-(4-Fluorophenyl)-5-methyl-1,2,3-triazole-4-carboxylic Acid

  • MDL number MFCD07397776
  • Cas Number : 887035-89-4
  • Molecular Weight : 221.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014319&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY014319.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart