AC5372911109 Accela ChemBio Inc.

3-(Boc-amino)-3-(4-chlorophenyl)propionic Acid

  • MDL number MFCD02090708
  • Cas Number : 284493-65-8
  • Molecular Weight : 299.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014450&attname=GoodsCode

Safety Information

signalword Warning
pcodes P261-P302+P352-P280-P305+P351+P338
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY014450.pdf
hstatements H315- H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart