AC5372911114 Accela ChemBio Inc.

3-(Dimethylamino)-1-(2-thienyl)-1-propanol

  • MDL number MFCD09701963
  • Cas Number : 13636-02-7
  • Molecular Weight : 185.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014457&attname=GoodsCode

Safety Information

signalword Warning
pcodes P264-P280-P302+P350-P305+P351+P338-P332+P313-P337+P313-P362
hcodes GHS07,
MSDS http://pdf.accelachem.com/cn/msds/SY014457.pdf
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart