AC5372911126 Accela ChemBio Inc.

1,2-Dihydro-3H-1,2,4-triazole-3-one

  • MDL number MFCD01311031
  • Cas Number : 930-33-6
  • Molecular Weight : 85.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014481&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY014481.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart