AC5372911134 Accela ChemBio Inc.

2-(Boc-amino)-2-(3-pyridinyl)acetic Acid

  • MDL number MFCD02683173
  • Cas Number : 347187-29-5
  • Molecular Weight : 252.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014512&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY014512.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart