AC5372911147 Accela ChemBio Inc.

3-Methyl-1-(2-pyridyl)-1-butylamine

  • MDL number MFCD12153903
  • Cas Number : 825647-69-6
  • Molecular Weight : 164.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014557&attname=GoodsCode

Safety Information

signalword Warning
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY014557.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart