AC5372911148 Accela ChemBio Inc.

1-Cyclopropyl-1-(2-pyridyl)methylamine

  • MDL number MFCD05191946
  • Cas Number : 535925-68-9
  • Molecular Weight : 148.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014560&attname=GoodsCode

Safety Information

signalword Warning
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY014560.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart