AC5372911149 Accela ChemBio Inc.

1-Cyclopropyl-1-(3-pyridyl)methylamine

  • MDL number MFCD05191947
  • Cas Number : 535925-69-0
  • Molecular Weight : 148.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014562&attname=GoodsCode

Safety Information

signalword Danger
pcodes P280;P305+P351+P338;P310
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY014562.pdf
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart