AC5372911155 Accela ChemBio Inc.

Methyl 7-Nitro-1,4-benzodioxane-6-carboxylate

  • MDL number MFCD18432414
  • Cas Number : 52791-03-4
  • Molecular Weight : 239.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014620&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY014620.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart