AC5372911180 Accela ChemBio Inc.

(R)-4,4,4-Trifluoro-3-hydroxybutanoic Acid

  • MDL number MFCD03093017
  • Cas Number : 108211-36-5
  • Molecular Weight : 158.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014726&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY014726.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart