AC5372911183 Accela ChemBio Inc.

Isopropyl 3-Aminocrotonate

  • MDL number MFCD00134272
  • Cas Number : 14205-46-0
  • Molecular Weight : 143.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014749&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY014749.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart