AC5372911184 Accela ChemBio Inc.

Ethyl 5-(Ethylsulfonyl)pyridine-2-carboxylate

  • MDL number MFCD19442771
  • Cas Number : 1314406-40-0
  • Molecular Weight : 243.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014754&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY014754.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart