AC5372911188 Accela ChemBio Inc.

Ethyl 4-Boc-2-morpholinecarboxylate

  • MDL number MFCD01646390
  • Cas Number : 768371-16-0
  • Molecular Weight : 259.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014774&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY014774.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart