AC5372911192 Accela ChemBio Inc.

Methyl (E)-2-Cyano-3-phenylacrylate

  • MDL number MFCD00025864
  • Cas Number : 14533-86-9
  • Molecular Weight : 187.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014783&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY014783.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart