AC5372911223 Accela ChemBio Inc.

(S)-(+)-2-(2-Chlorophenyl)glycine Methyl Ester Tartrate

  • MDL number MFCD15071471
  • Molecular Weight : 349.72104

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015015&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015015.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart