AC5372911235 Accela ChemBio Inc.

1-(3-Chlorophenyl)-4-oxocyclohexanecarboxylic Acid

  • MDL number MFCD06410577
  • Cas Number : 887978-71-4
  • Molecular Weight : 252.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015086&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015086.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart