AC5372911251 Accela ChemBio Inc.

1-(Boc-amino)-3-(methylamino)propane Hydrochloride

  • MDL number MFCD11101326
  • Cas Number : 1188264-02-9
  • Molecular Weight : 224.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015196&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015196.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart