AC5372911258 Accela ChemBio Inc.

Ethyl 2-Fluoro-6-nitrobenzoate

  • MDL number MFCD10566814
  • Cas Number : 1154426-16-0
  • Molecular Weight : 213.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015284&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015284.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart