AC5372911268 Accela ChemBio Inc.

Ethyl Pyridazine-4-carboxylate

  • MDL number MFCD09833000
  • Cas Number : 39123-39-2
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015372&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015372.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart