AC5372911279 Accela ChemBio Inc.

(8R,9S,10R,13S,14S)-10,13-Dimethylspiro[7,8,9,11,12,13,15,16-octahydro-6H-cyclopenta[a]phenanthrene-6,2'-oxirane]-3,17(10H,14H)-dione

  • MDL number MFCD19442795
  • Cas Number : 184972-12-1
  • Molecular Weight : 312.40276

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015402&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY015402.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart