AC5372911280 Accela ChemBio Inc.

Ethyl 3-Amino-3-(3-pyridyl)propionate Dihydrochloride

  • MDL number MFCD11053562
  • Cas Number : 149498-96-4
  • Molecular Weight : 267.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015408&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015408.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart