AC5372911283 Accela ChemBio Inc.

Methyl 3-Amino-2,2-dibenzylpropanoate

  • MDL number MFCD19442796
  • Cas Number : 125469-89-8
  • Molecular Weight : 283.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015414&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015414.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart