AC5372911294 Accela ChemBio Inc.

2-Methoxy-3-(trifluoromethyl)pyridine-5-boronic Acid Pinacol Ester

  • MDL number MFCD12026094
  • Cas Number : 1150561-61-7
  • Molecular Weight : 303.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015486&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015486.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart