AC5372911300 Accela ChemBio Inc.

4-Amino-3,5-difluorophenol

  • MDL number MFCD03094491
  • Cas Number : 135086-76-9
  • Molecular Weight : 145.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. -20?
Physical Form solid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015540&attname=GoodsCode

Safety Information

signalword Warning
pcodes P280H-P305+P351+P338
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY015540.pdf
hstatements H302-H312-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart