AC5372911303 Accela ChemBio Inc.

trans-2-Bromo-1-indanol

  • MDL number MFCD00151318
  • Cas Number : 10368-44-2
  • Molecular Weight : 213.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015553&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015553.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart