AC5372911308 Accela ChemBio Inc.

6-(3-Methoxyphenyl)nicotinic Acid

  • MDL number MFCD06410401
  • Cas Number : 887976-16-1
  • Molecular Weight : 229.23134

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015584&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015584.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart