AC5372911309 Accela ChemBio Inc.

3-(Boc-amino)-3-(4-tetrahydropyranyl)propanoic Acid

  • MDL number MFCD04115541
  • Cas Number : 887588-90-1
  • Molecular Weight : 273.32542

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015607&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015607.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart