AC5372911310 Accela ChemBio Inc.

2-Amino-2-(3-tetrahydrofuranyl)ethanol

  • MDL number MFCD04115614
  • Cas Number : 889949-68-2
  • Molecular Weight : 131.17292

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015608&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015608.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart